CAS RN 1797008-83-3
CAS RN 1797008-83-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-(2-Cyclohexyl-2-hydroxy-2-phenylacetyloxy)acetic Acid, TRC
CAS: 1797008-83-3 Molecular Formula: C16H20O5 Molecular Weight (g/mol): 292.33 IUPAC Name: 2-(2-cyclohexyl-2-hydroxy-2-phenylacetoxy)acetic acid SMILES: OC(COC(C(C1CCCCC1)(O)C2=CC=CC=C2)=O)=O