CAS RN 1797883-74-9
CAS RN 1797883-74-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-(2-Fluoro[1,1'-biphenyl]-4-yl)-2,3-dimethylbutanedioic Acid, TRC
CAS: 1797883-74-9 Molecular Formula: C18 H17 F O4 Molecular Weight (g/mol): 316.32 Synonym: Flurbiprofen EP Impurity B IUPAC Name: 2-(3-fluoro-4-phenyl-phenyl)-2,3-dimethyl-butanedioic acid SMILES: CC(C(=O)O)C(C)(C(=O)O)c1ccc(c(F)c1)c2ccccc2