accessibility menu, dialog, popup

UserName

CAS RN 1798-11-4

HOOONOO

CAS RN 1798-11-4

IUPAC Name: 2-(4-nitrophenoxy)acetic acid
Synonyms: P-nitrophenoxyacetic acid
Molecular Weight (g/mol): 197.15
Molecular Formula: C8H7NO5
InChi Key: AVDLFIONKHGQAP-UHFFFAOYSA-N
SMILES: OC(=O)COC1=CC=C(C=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results