accessibility menu, dialog, popup

UserName

CAS RN 179898-34-1

FBrN

CAS RN 179898-34-1

IUPAC Name: 3-bromo-5-fluorobenzonitrile
Synonyms: 3-bromo-5-fluorobenzonitrile
Molecular Weight (g/mol): 200.01
Molecular Formula: C7H3BrFN
InChi Key: IADLVSLZPQYXIF-UHFFFAOYSA-N
SMILES: FC1=CC(Br)=CC(=C1)C#N
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results