CAS RN 1802251-00-8
CAS RN 1802251-00-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
GSK1379725A, MedChemExpress
MedChemExpress GSK1379725A is a selective BPTF ligand with a Kd of 2.8 uM, showing no binding activity for Brd4.