accessibility menu, dialog, popup

UserName

CAS RN 1813-33-8

FFFClN

CAS RN 1813-33-8

IUPAC Name: 2-chloro-4-(trifluoromethyl)benzonitrile
Synonyms: 2-chloro-4-(trifluoromethyl)benzonitrile
Molecular Weight (g/mol): 205.56
Molecular Formula: C8H3ClF3N
InChi Key: GEHMLBFNZKJDQM-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(Cl)=C(C=C1)C#N
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results