accessibility menu, dialog, popup

UserName

CAS RN 1824186-34-6

NClNHSNH2

CAS RN 1824186-34-6

IUPAC Name: (4-chloropyridin-2-yl)thiourea
Synonyms: 4-chloropyridin-2-ylthiourea
Molecular Weight (g/mol): 187.65
Molecular Formula: C6H6ClN3S
InChi Key: MWEAHCQSCSTMKP-UHFFFAOYSA-N
SMILES: NC(=S)NC1=NC=CC(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results