accessibility menu, dialog, popup

UserName

HNOHNNOOHOBr

CAS RN 182498-32-4

IUPAC Name: 3-(2-bromophenyl)-1-(2-hydroxy-4-nitrophenyl)urea
Synonyms: 3-(2-bromophenyl)-1-(2-hydroxy-4-nitrophenyl)urea
Molecular Weight (g/mol): 352.14
Molecular Formula: C13H10BrN3O4
InChi Key: MQBZVUNNWUIPMK-UHFFFAOYSA-N
SMILES: OC1=C(NC(=O)NC2=CC=CC=C2Br)C=CC(=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (1)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
  • (5)
  • (5)
  • (2)