accessibility menu, dialog, popup

UserName

CAS RN 18303-04-3

IUPAC Name: tert-butyl N-(4-methylbenzenesulfonyl)carbamate
Synonyms: tert-butyl N-(4-methylbenzenesulfonyl)carbamate
Molecular Weight (g/mol): 271.33
Molecular Formula: C12H17NO4S
InChi Key: DUTLOVSBVBGNDM-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)NC(=O)OC(C)(C)C
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)