accessibility menu, dialog, popup

UserName

CAS RN 18511-62-1

OF

CAS RN 18511-62-1

IUPAC Name: 2-(4-fluorophenyl)oxirane
Synonyms: 2-(4-fluorophenyl)oxirane
Molecular Weight (g/mol): 138.14
Molecular Formula: C8H7FO
InChi Key: ICVNPQMUUHPPOK-UHFFFAOYNA-N
SMILES: FC1=CC=C(C=C1)C1CO1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results