CAS RN 18550-98-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3PO, MedChemExpress
MedChemExpress 3PO is a inhibitor of (PFKFB3). 3PO attenuates the proliferation of several cancer cell lines with IC50s of 1.4-24 μmol/L. 3PO suppresses glucose uptake and decreases the intracellular concentra tion of Fru-2,6-BP, lactate, ATP, NAD+, and NADH. 3PO can be used for the research of cancer.