accessibility menu, dialog, popup

UserName

CH3ONHOHOFOHH3CCH3H3C

CAS RN 185629-22-5

IUPAC Name: 3-fluoro-4-[2-hydroxy-2-(5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl)acetamido]benzoic acid
Synonyms: 3-fluoro-4-[2-hydroxy-2-(5,5,8,8-tetramethyl-6,7-dihydronaphthalen-2-yl)acetamido]benzoic acid
Molecular Weight (g/mol): 399.46
Molecular Formula: C23H26FNO4
InChi Key: AANFHDFOMFRLLR-UHFFFAOYNA-N
SMILES: CC1(C)CCC(C)(C)C2=C1C=CC(=C2)C(O)C(=O)NC1=C(F)C=C(C=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results