accessibility menu, dialog, popup

UserName

OOBr

CAS RN 18698-97-0

IUPAC Name: 2-(2-bromophenyl)acetate
Synonyms: 2-(2-bromophenyl)acetate
Molecular Weight (g/mol): 214.04
Molecular Formula: C8H6BrO2
InChi Key: DWXSYDKEWORWBT-UHFFFAOYSA-M
SMILES: [O-]C(=O)CC1=CC=CC=C1Br
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)