accessibility menu, dialog, popup

UserName

ClSOON

CAS RN 18704-37-5

IUPAC Name: quinoline-8-sulfonyl chloride
Synonyms: quinoline-8-sulfonyl chloride
Molecular Weight (g/mol): 227.66
Molecular Formula: C9H6ClNO2S
InChi Key: JUYUYCIJACTHMK-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=C2N=CC=CC2=CC=C1
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)