CAS RN 1871-89-2
CAS RN 1871-89-2
IUPAC Name:
N,N'-bis(2-hydroxyethyl)ethanediamide
Synonyms:
N,N'-bis(2-hydroxyethyl)ethanediamide
Molecular Weight (g/mol):
176.17
Molecular Formula:
C6H12N2O4
InChi Key:
FPQJEXTVQZHURJ-UHFFFAOYSA-N
SMILES:
OCCNC(=O)C(=O)NCCO
1
–
1
of
1
results
1
N.N'-Bis(2-hydroxyethyl)oxamide, 99%
CAS: 1871-89-2 Molecular Formula: C6H12N2O4 Molecular Weight (g/mol): 176.172 MDL Number: MFCD00020559 InChI Key: FPQJEXTVQZHURJ-UHFFFAOYSA-N Synonym: n,n'-bis 2-hydroxyethyl oxamide,n1,n2-bis 2-hydroxyethyl oxalamide,nn'-bis 2-hydroxyethyl oxamide,unii-t92n2yr78x,ethanediamide, n,n'-bis 2-hydroxyethyl,n,n-bis 2-hydroxyethyl oxamide,n,n'-bis-2-hydroxyethyl-oxamide,ethanediamide, n1,n2-bis 2-hydroxyethyl,n,n'-bis 2-hydroxyethyl ethanediamide,n-2-hydroxyethyl-n'-2-hydroxyethyl ethane-1,2-diamide PubChem CID: 74638 IUPAC Name: N,N'-bis(2-hydroxyethyl)oxamide SMILES: C(CO)NC(=O)C(=O)NCCO