accessibility menu, dialog, popup

UserName

CAS RN 1875-48-5

H2NNOO

CAS RN 1875-48-5

IUPAC Name: 2-amino-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms: 2-aminoisoindole-1,3-dione
Molecular Weight (g/mol): 162.15
Molecular Formula: C8H6N2O2
InChi Key: KSILMCDYDAKOJD-UHFFFAOYSA-N
SMILES: NN1C(=O)C2=CC=CC=C2C1=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results