CAS RN 18838-10-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-(2-hydroxyethyl)benzamide, TRC
CAS: 18838-10-3 Molecular Formula: C9H11NO2 Molecular Weight (g/mol): 165.18 IUPAC Name: N-(2-hydroxyethyl)benzamide SMILES: OCCNC(=O)c1ccccc1