CAS RN 188854-42-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1-[(Diethylamino)carbonyl]piperidine-4-carboxylic Acid, TRC
CAS: 188854-42-4 Molecular Formula: C11H20N2O3 Molecular Weight (g/mol): 228.29 Synonym: 1-[(Diethylamino)carbonyl]-4-piperidinecarboxylic Acid,1-Diethylcarbamoyl-piperidine-4-carboxylic Acid IUPAC Name: 1-(diethylcarbamoyl)piperidine-4-carboxylic acid SMILES: CCN(CC)C(=O)N1CCC(CC1)C(=O)O