accessibility menu, dialog, popup

UserName

CH3SnH3CCH3NBr

CAS RN 189083-81-6

IUPAC Name: 2-bromo-6-(tributylstannyl)pyridine
Synonyms: 2-bromo-6-(tributylstannyl)pyridine
Molecular Weight (g/mol): 447.05
Molecular Formula: C17H30BrNSn
InChi Key: ZRPIKMNUVOBWBE-UHFFFAOYSA-N
SMILES: CCCC[Sn](CCCC)(CCCC)C1=CC=CC(Br)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results