CAS RN 1899-93-0
IUPAC Name:
3-methylbenzene-1-sulfonyl chloride
Synonyms:
3-methylbenzenesulfonyl chloride
Molecular Weight (g/mol):
190.64
Molecular Formula:
C7H7ClO2S
InChi Key:
KFPMLWUKHQMEBU-UHFFFAOYSA-N
SMILES:
CC1=CC(=CC=C1)S(Cl)(=O)=O
1
–
2
of
2
results
1
m-Toluenesulfonyl chloride, 98%
CAS: 1899-93-0 Molecular Formula: C7H7ClO2S Molecular Weight (g/mol): 190.641 MDL Number: MFCD00051747 InChI Key: KFPMLWUKHQMEBU-UHFFFAOYSA-N Synonym: m-toluenesulfonyl chloride,3-methylbenzene-1-sulfonyl chloride,3-toluenesulfonyl chloride,3-methyl-benzenesulfonyl chloride,3-methylbenzenesulphonyl chloride,benzenesulfonyl chloride, 3-methyl,3-toluenesulphonyl chloride,3-methylphenylsulfonyl chloride,pubchem5624,m-tolylsulfonyl chloride PubChem CID: 2734595 IUPAC Name: 3-methylbenzenesulfonyl chloride SMILES: CC1=CC(=CC=C1)S(=O)(=O)Cl
2
m-Toluenesulfonyl Chloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.