accessibility menu, dialog, popup

UserName

CAS RN 18999-45-6

IUPAC Name: 3-(1H-imidazol-1-yl)propanoic acid
Synonyms: imidazolepropionic acid
Molecular Weight (g/mol): 140.14
Molecular Formula: C6H8N2O2
InChi Key: VSFNAZLYGOOSEY-UHFFFAOYSA-N
SMILES: OC(=O)CCN1C=CN=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results