accessibility menu, dialog, popup

UserName

CAS RN 19013-10-6

H3COOOHNOO

CAS RN 19013-10-6

IUPAC Name: ethyl 4-hydroxy-3-nitrobenzoate
Synonyms: ethyl 4-hydroxy-3-nitrobenzoate
Molecular Weight (g/mol): 211.17
Molecular Formula: C9H9NO5
InChi Key: FBHJNBWUGONVNS-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=C(O)C(=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results