accessibility menu, dialog, popup

UserName

ClONHOOHClH2O

CAS RN 190908-40-8

IUPAC Name: 5,7-dichloro-4-oxo-1,4-dihydroquinoline-2-carboxylic acid hydrate
Synonyms: 5,7-dichlorokynurenic acid hydrate
Molecular Weight (g/mol): 276.07
Molecular Formula: C10H7Cl2NO4
InChi Key: READYYZXDSXRQJ-UHFFFAOYSA-N
SMILES: O.OC(=O)C1=CC(=O)C2=C(Cl)C=C(Cl)C=C2N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results