accessibility menu, dialog, popup

UserName

CH3H2NNOH(E)

CAS RN 19227-13-5

IUPAC Name: (E)-N'-hydroxy-4-methylbenzene-1-carboximidamide
Synonyms: (E)-N'-hydroxy-4-methylbenzenecarboximidamide
Molecular Weight (g/mol): 150.18
Molecular Formula: C8H10N2O
InChi Key: NKJXMLIWSJATEE-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C(\N)=N/O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results