CAS RN 19244-98-5
IUPAC Name:
hydrogen guanidine bromide
Synonyms:
hydrogen guanidine bromide
Molecular Weight (g/mol):
139.98
Molecular Formula:
CH6BrN3
InChi Key:
VQNVZLDDLJBKNS-UHFFFAOYSA-N
SMILES:
[H+].[Br-].NC(N)=N