accessibility menu, dialog, popup

UserName

BNOOFFFK

CAS RN 192863-40-4

IUPAC Name: potassium trifluoro(3-nitrophenyl)boranuide
Synonyms: potassium trifluoro(3-nitrophenyl)boranuide
Molecular Weight (g/mol): 229.01
Molecular Formula: C6H4BF3KNO2
InChi Key: ZKZMSCLFLMORNI-UHFFFAOYSA-N
SMILES: [K+].[O-][N+](=O)C1=CC=CC(=C1)[B-](F)(F)F
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)