CAS RN 193273-69-7
CAS RN 193273-69-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Capromorelin Tartrate, MedChemExpress
MedChemExpress Capromorelin Tartrate is an orally active, potent growth hormone secretagogue receptor (GHSR) agonist, with Ki of 7 nM for hGHS-R1a.