accessibility menu, dialog, popup

UserName

ClClO

CAS RN 19347-73-0

IUPAC Name: 6-chlorohexanoyl chloride
Synonyms: 6-chlorohexanoyl chloride
Molecular Weight (g/mol): 169.05
Molecular Formula: C6H10Cl2O
InChi Key: WZILXAPNPKMOSA-UHFFFAOYSA-N
SMILES: ClCCCCCC(Cl)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results