accessibility menu, dialog, popup

UserName

CAS RN 19395-87-0

H3COHNNHONClO

CAS RN 19395-87-0

IUPAC Name: N'-acetyl-13-chloro-2-oxa-9-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaene-9-carbohydrazide
Synonyms: N'-acetyl-13-chloro-2-oxa-9-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaene-9-carbohydrazide
Molecular Weight (g/mol): 331.76
Molecular Formula: C16H14ClN3O3
InChi Key: UETSVHKFTALXNP-UHFFFAOYSA-N
SMILES: CC(=O)NNC(=O)N1CC2=CC(Cl)=CC=C2OC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results