accessibility menu, dialog, popup

UserName

CAS RN 1942-45-6

CH3H3C

CAS RN 1942-45-6

IUPAC Name: oct-4-yne
Synonyms: 4-octyne
Molecular Weight (g/mol): 110.2
Molecular Formula: C8H14
InChi Key: GZTNBKQTTZSQNS-UHFFFAOYSA-N
SMILES: CCCC#CCCC
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)