CAS RN 194209-18-2
CAS RN 194209-18-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1-(1-methyl-1H-1,2,3-triazol-5-yl)ethan-1-one, TRC
CAS: 194209-18-2 Molecular Formula: C5H7N3O Molecular Weight (g/mol): 125.13 IUPAC Name: 1-(3-methyltriazol-4-yl)ethanone SMILES: CC(=O)c1cnnn1C