accessibility menu, dialog, popup

UserName

CH3NH2NNNHCl(E/Z)

CAS RN 194852-88-5

IUPAC Name: N'-methyl-1H-pyrazole-1-carboximidamide hydrochloride
Synonyms: N'-methylpyrazole-1-carboximidamide hydrochloride
Molecular Weight (g/mol): 160.61
Molecular Formula: C5H9ClN4
InChi Key: SYSWHVMZAHBYJO-UHFFFAOYSA-N
SMILES: Cl.CN=C(N)N1C=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results