accessibility menu, dialog, popup

UserName

CAS RN 1949-89-9

(R)O(R)(R)OHHOHOHO

CAS RN 1949-89-9

IUPAC Name: (3R,4R,5R)-3,4,5,6-tetrahydroxyhexanal
Synonyms: 2-deoxy-D-galactose 2-deoxy-D-lyxo-hexose
Molecular Weight (g/mol): 164.16
Molecular Formula: C6H12O5
InChi Key: VRYALKFFQXWPIH-HSUXUTPPSA-N
SMILES: OC[C@@H](O)[C@H](O)[C@H](O)CC=O
Quantity 
  • (2)
  • (1)
  • (1)
  • (4)
Physical Form 
  • (2)
  • (4)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (4)
  • (2)
  • (4)
  • (1)