CAS RN 1950-85-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Sodium Methanethiosulfonate, TRC
CAS: 1950-85-2 Molecular Formula: C H3 Na O2 S2 Molecular Weight (g/mol): 134.15 IUPAC Name: methylsulfonylsulfanylsodium SMILES: CS(=O)(=O)S[Na]