accessibility menu, dialog, popup

UserName

CAS RN 19562-30-2

IUPAC Name: 8-ethyl-5-oxo-2-(pyrrolidin-1-yl)-5H,8H-pyrido[2,3-d]pyrimidine-6-carboxylic acid
Synonyms: enterol acidum piromidicum septural acid, piromidic acide piromidique uropir zaomeal piromidic acid pirodal urisept
Molecular Weight (g/mol): 288.31
Molecular Formula: C14H16N4O3
InChi Key: RCIMBBZXSXFZBV-UHFFFAOYSA-N
SMILES: CCN1C=C(C(O)=O)C(=O)C2=CN=C(N=C12)N1CCCC1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Color 
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results