CAS RN 195875-84-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tesofensine, TRC
CAS: 195875-84-4 Molecular Formula: C17 H23 Cl2 N O Molecular Weight (g/mol): 328.28 Synonym: (1R,2R,3S,5S)-3-(3,4-Dichlorophenyl)-2-(ethoxymethyl)-8-methyl-8-azabicyclo[3.2.1]octane,[1R-(2-endo,3-exo)]-3-(3,4-dichlorophenyl)-2-(ethoxymethyl)-8-methyl-8-azabicyclo[3.2.1]octane,NS 2330; IUPAC Name: (1S,3S,4R,5R)-3-(3,4-dichlorophenyl)-4-(ethoxymethyl)-8-methyl-8-azabicyclo[3.2.1]octane SMILES: CCOC[C@H]1[C@H]2CC[C@@H](C[C@@H]1c3ccc(Cl)c(Cl)c3)N2C