accessibility menu, dialog, popup

UserName

BOOCH3CH3CH3CH3H3CCH3BOOH3CH3CH3CH3C

CAS RN 196207-58-6

IUPAC Name: 2-[9,9-dioctyl-7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-fluoren-2-yl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-[9,9-dioctyl-7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)fluoren-2-yl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 642.58
Molecular Formula: C41H64B2O4
InChi Key: FAHIZHKRQQNPLC-UHFFFAOYSA-N
SMILES: CCCCCCCCC1(CCCCCCCC)C2=C(C=CC(=C2)B2OC(C)(C)C(C)(C)O2)C2=C1C=C(C=C2)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results