CAS RN 196808-24-9
CAS RN 196808-24-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
GW1929, MedChemExpress
MedChemExpress GW1929 is a potent PPAR-γ agonist, with a pKi of 8.84 for human PPAR-γ, and pEC50s of 8.56 and 8.27 for human PPAR-γ and murine PPAR-γ, respectively.