accessibility menu, dialog, popup

UserName

CH3NNCH3Br(E/Z)(E/Z)

CAS RN 19764-88-6

IUPAC Name: 1-ethyl-4-[3-(1-ethyl-1,4-dihydroquinolin-4-ylidene)prop-1-en-1-yl]quinolin-1-ium bromide
Synonyms: cryptocyanin cation bromide
Molecular Weight (g/mol): 433.39
Molecular Formula: C25H25BrN2
InChi Key: QRRYUUHAKZXUIQ-UHFFFAOYSA-M
SMILES: [Br-].CCN1C=CC(=CC=CC2=C3C=CC=CC3=[N+](CC)C=C2)C2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results