accessibility menu, dialog, popup

UserName

CH3SOOHNN(E/Z)

CAS RN 19816-85-4

IUPAC Name: N'-(1,2-diphenylethylidene)-4-methylbenzene-1-sulfonohydrazide
Synonyms: N'-(1,2-diphenylethylidene)-4-methylbenzenesulfonohydrazide
Molecular Weight (g/mol): 364.46
Molecular Formula: C21H20N2O2S
InChi Key: INMYHVHTZKDKDK-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)NN=C(CC1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results