accessibility menu, dialog, popup

UserName

H3CCH3H3COOHNOHONHOO

CAS RN 198544-42-2

IUPAC Name: 3-{[(tert-butoxy)carbonyl]amino}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
Synonyms: 3-[(tert-butoxycarbonyl)amino]-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoic acid
Molecular Weight (g/mol): 426.47
Molecular Formula: C23H26N2O6
InChi Key: PKAUMAVONPSDRW-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results