accessibility menu, dialog, popup

UserName

OHO(S)HCNHOO

CAS RN 198561-07-8

IUPAC Name: (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)pent-4-ynoic acid
Synonyms: (2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}pent-4-ynoic acid
Molecular Weight (g/mol): 335.36
Molecular Formula: C20H17NO4
InChi Key: DJGMNCKHNMRKFM-SFHVURJKSA-N
SMILES: OC(=O)[C@H](CC#C)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)