accessibility menu, dialog, popup

UserName

CAS RN 199666-03-0

CH3OOCH3SOOHNSNNOO

CAS RN 199666-03-0

IUPAC Name: 3,4-dimethoxy-N-[4-(3-nitrophenyl)-1,3-thiazol-2-yl]benzene-1-sulfonamide
Synonyms: 3,4-dimethoxy-N-[4-(3-nitrophenyl)-1,3-thiazol-2-yl]benzenesulfonamide
Molecular Weight (g/mol): 421.44
Molecular Formula: C17H15N3O6S2
InChi Key: NDPBMCKQJOZAQX-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1OC)S(=O)(=O)NC1=NC(=CS1)C1=CC=CC(=C1)[N+]([O-])=O
Grade 
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results