accessibility menu, dialog, popup

UserName

CAS RN 199915-38-3

OONCS

CAS RN 199915-38-3

IUPAC Name: (9H-fluoren-9-yl)methyl N-carbothioylcarbamate
Synonyms: 9H-fluoren-9-ylmethyl N-carbothioylcarbamate
Molecular Weight (g/mol): 281.33
Molecular Formula: C16H11NO2S
InChi Key: DHMYULZVFHHEHE-UHFFFAOYSA-N
SMILES: O=C(OCC1C2=CC=CC=C2C2=CC=CC=C12)N=C=S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results