accessibility menu, dialog, popup

UserName

CAS RN 200573-11-1

CH3OOOSOOFFF

CAS RN 200573-11-1

IUPAC Name: methyl 2-(trifluoromethanesulfonyloxy)naphthalene-1-carboxylate
Synonyms: methyl 2-(trifluoromethanesulfonyloxy)naphthalene-1-carboxylate
Molecular Weight (g/mol): 334.27
Molecular Formula: C13H9F3O5S
InChi Key: BDXDWDCWSCPFEW-UHFFFAOYSA-N
SMILES: COC(=O)C1=C2C=CC=CC2=CC=C1OS(=O)(=O)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results