accessibility menu, dialog, popup

UserName

CAS RN 201484-50-6

IUPAC Name: (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3,3-diphenylpropanoic acid
Synonyms: (2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3,3-diphenylpropanoic acid
Molecular Weight (g/mol): 463.53
Molecular Formula: C30H25NO4
InChi Key: PENQOTJCVODUQU-NDEPHWFRSA-N
SMILES: OC(=O)[C@@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)