accessibility menu, dialog, popup

UserName

NH2PHOHOOOHO

CAS RN 20263-07-4

IUPAC Name: 2-amino-4-phosphonobutanoic acid
Synonyms: 2-amino-4-phosphonobutanoic acid
Molecular Weight (g/mol): 183.1
Molecular Formula: C4H10NO5P
InChi Key: DDOQBQRIEWHWBT-UHFFFAOYNA-N
SMILES: NC(CCP(O)(O)=O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results