accessibility menu, dialog, popup

UserName

H3CONNOO

CAS RN 20265-37-6

IUPAC Name: 3-methoxy-2-nitropyridine
Synonyms: 3-methoxy-2-nitropyridine
Molecular Weight (g/mol): 154.13
Molecular Formula: C6H6N2O3
InChi Key: LSXHCFSGOBFNDX-UHFFFAOYSA-N
SMILES: COC1=C(N=CC=C1)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)