accessibility menu, dialog, popup

UserName

CAS RN 202821-81-6

NNOOClOHFFF

CAS RN 202821-81-6

IUPAC Name: 3-[(5-chloro-2-hydroxyphenyl)methyl]-5-[4-(trifluoromethyl)phenyl]-2,3-dihydro-1,3,4-oxadiazol-2-one
Synonyms: 3-[(5-chloro-2-hydroxyphenyl)methyl]-5-[4-(trifluoromethyl)phenyl]-1,3,4-oxadiazol-2-one
Molecular Weight (g/mol): 370.71
Molecular Formula: C16H10ClF3N2O3
InChi Key: QKOWACXSXTXRKA-UHFFFAOYSA-N
SMILES: OC1=C(CN2N=C(OC2=O)C2=CC=C(C=C2)C(F)(F)F)C=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results