accessibility menu, dialog, popup

UserName

H3NBrF

CAS RN 202865-69-8

IUPAC Name: (5-bromo-2-fluorophenyl)methanaminium
Synonyms: (5-bromo-2-fluorophenyl)methanaminium
Molecular Weight (g/mol): 205.05
Molecular Formula: C7H8BrFN
InChi Key: PDKBJZGGXHTHNC-UHFFFAOYSA-O
SMILES: [NH3+]CC1=CC(Br)=CC=C1F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results